Tisk…

2,3-Dimethoxybenzoic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

D131407-25G D131407-5G
SIALD131407-25GEA 2410 CZK
SIALD131407-25G SIALD131407-5G
2,3-Dimethoxybenzoic acid, Sigma-Aldrich®
2,3-Dimethoxybenzoic acid
Vzorec: C₉H₁₀O₄
MW: 182,18 g/mol
Bod varu: 314 °C (1013 hPa)
Bod tání: 121…124 °C
Hustota: 1,387 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002432
CAS číslo: 1521-38-6
EINECS: 216-188-4

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR