o-Anisic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
169978-100G
169978-25G
SIAL169978-100GEA
1720
CZK
SIAL169978-100G
SIAL169978-25G
o-Anisic acid, Sigma-Aldrich®
o-Anisic acid
Vzorec:
C₈H₈O₃ Bod varu: 280 °C (1013 hPa) Bod tání: 99 °C Hustota: 1,38 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002431 CAS číslo: 579-75-9 EINECS: 209-447-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...