Tisk…

4,5-Dimethoxy-2-nitrobenzoic acid, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

329231-100G
SIAL329231-100GEA 7760 CZK
SIAL329231-100G
4,5-Dimethoxy-2-nitrobenzoic acid, Sigma-Aldrich®
4,5-Dimethoxy-2-nitrobenzoic acid
Vzorec: C₉H₉NO₆
MW: 227.17
MDL Number: MFCD00014697
CAS číslo: 4998-07-6
EINECS: 225-657-2

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR