4-(Methylamino)benzoic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
119695-10G
119695-50G
SIAL119695-10GEA
1990
CZK
SIAL119695-10G
SIAL119695-50G
4-(Methylamino)benzoic acid, Sigma-Aldrich®
4-(Methylamino)benzoic acid
Vzorec:
C₈H₉NO₂ MW: 151,16 g/mol Bod varu: 296 °C (1013 hPa) Bod tání: 159 °C |
MDL Number:
MFCD00002535 CAS číslo: 10541-83-0 EINECS: 234-121-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...