3-Aminosalicylic acid, powder
Dodavatel: MP Biomedicals
|
Varování
|
Synonyms:
3-Aminosalicylic Acid, 3-Amino-2-hydroxybenzoic acid, 3-Amino-2-hydroxybenzoic acid
0215476990
0215476980
ICNA0215476990EA
9530
CZK
ICNA0215476990
ICNA0215476980
3-Aminosalicylic acid, powder
3-Aminosalicylic acid
Soluble in KOH, clear solution.
Vzorec:
C₇H₇NO₃ MW: 153,14 g/mol Bod varu: 347 °C (1013 hPa) Bod tání: 290 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010299 CAS číslo: 570-23-0 EINECS: 209-328-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...