Tisk…

4-Amino-m-anisic acid 95%

Dodavatel: Thermo Fisher Scientific
Varování

H28098.06 H28098.03
H28098.06EA 8100 CZK
H28098.06 H28098.03
4-Amino-m-anisic acid 95%
4-Amino-m-anisic acid

Purity: 95% CAS No.: 2486-69-3 Molecular Formula: C8H9NO3 Formula Weight: 167.16 Melting Point: 186-188° MDL No.: MFCD00016539

Vzorec: C₈H₉NO₃
Bod tání: 186…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00016539
CAS číslo: 2486-69-3

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR