5-Amino-m-anisic acid, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
694428-1G
694428-5G
SIAL694428-1GEA
1450
CZK
SIAL694428-1G
SIAL694428-5G
5-Amino-m-anisic acid, Sigma-Aldrich®
5-Amino-m-anisic acid
Vzorec:
C₈H₉NO₃ MW: 167,16 g/mol |
MDL Number:
MFCD07783718 CAS číslo: 74165-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...