Silver benzoate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Benzoic acid silver (I) salt
M-Clarity™ quality level = MQ200
227277-10G
227277-50G
SIAL227277-10GEA
2400
CZK
SIAL227277-10G
SIAL227277-50G
Silver benzoate, Sigma-Aldrich®
Silver benzoate
Vzorec:
C₇H₅AgO₂ MW: 228,98 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00013030 CAS číslo: 532-31-0 EINECS: 208-533-2 Merck Index: 13,01092 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...