Tisk…

2-Cyanobenzoic acid, Sigma-Aldrich®

Dodavatel: Merck

527742-1G 527742-5G
SIAL527742-1GEA 2010 CZK
SIAL527742-1G SIAL527742-5G
2-Cyanobenzoic acid, Sigma-Aldrich®
2-Cyanobenzoic acid
Vzorec: C₈H₅NO₂
MW: 147,13 g/mol
MDL Number: MFCD00045796
CAS číslo: 3839-22-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR