Tisk…

Dimethyl terephthalate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ200

185124-3KG 185124-500G
SIAL185124-3KGEA 3940 CZK
SIAL185124-3KG SIAL185124-500G
Dimethyl terephthalate, Sigma-Aldrich®
Dimethyl terephthalate
Vzorec: C₁₀H₁₀O₄
Bod varu: 288 °C (1013 hPa)
Bod tání: 139 °C
Hustota: 1,2 g/cm³ (20 °C)
Bod vzplanutí: 141 °C
Storage Temperature: Ambient
MDL Number: MFCD00008440
CAS číslo: 120-61-6
EINECS: 204-411-8
Merck Index: 12,09306

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR