Propyl 4-hydroxybenzoát, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester
M-Clarity™ quality level = MQ200
P5835-100G
P5835-500G
SIALP5835-100GEA
4460
CZK
SIALP5835-100G
SIALP5835-500G
Propyl 4-hydroxybenzoát, Sigma-Aldrich®
Propyl 4-hydroxybenzoát
Vzorec:
C₁₀H₁₂O₃ Bod varu: 292 °C (1013 hPa) Bod tání: 95 °C Hustota: 1,28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002354 CAS číslo: 94-13-3 EINECS: 202-307-7 Merck Index: 13,07958 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...