Tisk…

methyl 4-hydroxybenzoát, Sigma-Aldrich®

Dodavatel: Merck
Varování

Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate

M-Clarity™ quality level = MQ200

H5501-1KG H5501-500G H5501-5KG H5501-100G
SIALH5501-1KGEA 7620 CZK
SIALH5501-1KG SIALH5501-500G SIALH5501-5KG SIALH5501-100G
methyl 4-hydroxybenzoát, Sigma-Aldrich®
methyl 4-hydroxybenzoát
Vzorec: HOC₆H₄CO₂CH₃
MW: 152.15
Bod varu: 270…280 °C (1013 hPa)
Bod tání: 125…128 °C
Hustota: 1.1208 g/cm³ (137 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002352
CAS číslo: 99-76-3
EINECS: 202-785-7
Merck Index: 13,06129
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84321052052f4be2a53d8d7ebac7fabe6fc5a435&vid=ede765fad275b2515a8c57ba4499bff313a18983&prid=web&clid=SAPDAM

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR