Methyl-3-hydroxybenzoate, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
Methyl 3-Hydroxybenzoate, 3-Hydroxybenzoic acid methyl ester
M-Clarity™ quality level = MQ100
252794-100G
252794-25G
SIAL252794-100GEA
5970
CZK
SIAL252794-100G
SIAL252794-25G
Methyl-3-hydroxybenzoate, Sigma-Aldrich®
Methyl-3-hydroxybenzoate
Vzorec:
C₈H₈O₃ MW: 152,15 g/mol |
MDL Number:
MFCD00002295 CAS číslo: 19438-10-9 EINECS: 243-071-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...