Methyl-3-hydroxybenzoate ≥99%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
Methyl 3-Hydroxybenzoate, 3-Hydroxybenzoic acid methyl ester
A14425.14
A14425.22
A14425.36
A14425.14EA
1780
CZK
A14425.14
A14425.22
A14425.36
Methyl-3-hydroxybenzoate ≥99%
Methyl-3-hydroxybenzoate
Purity: 99%
CAS No.: 19438-10-9
Molecular Formula: C8H8O3
Formula Weight: 152.15
Melting Point: 70-72°
Boiling Point: 280-281°/708mm
MDL No.: MFCD00002295
Vzorec:
C₈H₈O₃ MW: 152.15 Bod varu: 280…281 °C (708 mmHg) Bod tání: 70…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002295 CAS číslo: 19438-10-9 EINECS: 243-071-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...