Tisk…

Ethyl-3-nitrobenzoate ≥98%

Dodavatel: Thermo Scientific

Synonyms: Ethyl 3-nitrobenzoate, 3-Nitrobenzoic acid ethyl ester

A10626.14 A10626.22
A10626.14EA 1580 CZK
A10626.14 A10626.22
Ethyl-3-nitrobenzoate ≥98%
Ethyl-3-nitrobenzoate

Purity: 98+% CAS No.: 618-98-4 Molecular Formula: C9H9NO4 Formula Weight: 195.18 Melting Point: 40-43° Boiling Point: 297-298° Merck Reference: 14,3832 MDL No.: MFCD00014702

Vzorec: C₉H₉NO₄
MW: 195,17 g/mol
Bod varu: 297…298 °C
Bod tání: 40…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00014702
CAS číslo: 618-98-4
EINECS: 210-574-6
Merck Index: 14,3832

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR