Ethyl-2-nitrobenzoate ≥97%
Dodavatel: Thermo Scientific
Synonyms:
Ethyl 2-nitrobenzoate, 2-Nitrobenzoic acid ethyl ester
A19994.14
A19994.22
A19994.14EA
2130
CZK
A19994.14
A19994.22
Ethyl-2-nitrobenzoate ≥97%
Ethyl-2-nitrobenzoate
Purity: 97%
CAS No.: 610-34-4
Molecular Formula: C9H9NO4
Formula Weight: 195.18
Melting Point: 26-30°
Boiling Point: 172-174°/18mm
Merck Reference: 14,3832
MDL No.: MFCD00014696
Vzorec:
C₉H₉NO₄ MW: 195,17 g/mol Bod varu: 172…174 °C (18 mmHg) Bod tání: 26…30 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014696 CAS číslo: 610-34-4 EINECS: 210-220-0 Merck Index: 14,3832 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...