Methyl-N-methylanthranilate, Sigma-Aldrich®
Dodavatel: Merck
|
Varování
|
Synonyms:
Methyl N-Methylanthranilate, Dimethyl anthranilate
M-Clarity™ quality level = MQ300
W271802-1KG-K
W271802-5KG-K
SIALW271802-1KG-KEA
7430
CZK
SIALW271802-1KG-K
SIALW271802-5KG-K
Methyl-N-methylanthranilate, Sigma-Aldrich®
Methyl-N-methylanthranilate
Vzorec:
C₉H₁₁NO₂ MW: 165.19 Hustota: 1.12 g/cm³ (15 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00017183 CAS číslo: 85-91-6 EINECS: 201-642-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...