Ethyl anthranilate ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Aminobenzoic acid ethyl ester, o-Aminobenzoic acid ethyl ester, Ethyl 2-aminobenzoate
A14378.18
A14378.22
A14378.18EA
975
CZK
A14378.18
A14378.22
Ethyl anthranilate ≥98%
Ethyl anthranilate
Purity: 98+%
CAS No.: 87-25-2
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Flash Point: 138°(280°F)
Physical Form: Liquid
Density: 1.118
Refractive Index: 1.5640
Appearance: Colourless to pale yellow
Odour: Aromatic
Melting Point: 14-15°
Boiling Point: 266-268°
Beilstein Reference: 878874
MDL No.: MFCD00007711
Vzorec:
C₉H₁₁NO₂ MW: 165,19 g/mol Bod varu: 266…268 °C Bod tání: 14…15 °C Hustota: 1,118 Bod vzplanutí: 138 °C (280 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007711 CAS číslo: 87-25-2 EINECS: 201-735-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...