Tisk…

Hexyl benzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: Benzoic acid hexyl ester

M-Clarity™ quality level = MQ300

W369101-1KG-K W369101-5KG-K
SIALW369101-1KG-KEA 0 CZK
SIALW369101-1KG-K SIALW369101-5KG-K
Hexyl benzoate, Sigma-Aldrich®
Hexyl benzoate
Vzorec: C₁₃H₁₈O₂
MW: 206,28 g/mol
Bod varu: 272 °C (1013 hPa)
Hustota: 0,9992 g/cm³ (0 °C)
MDL Number: MFCD00051714
CAS číslo: 6789-88-4
EINECS: 229-856-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR