Tisk…

Isopentyl benzoate, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ300

W205818-10KG-K W205818-1KG-K
SIALW205818-10KG-KEA 15690 CZK
SIALW205818-10KG-K SIALW205818-1KG-K
Isopentyl benzoate, Sigma-Aldrich®
Isopentyl benzoate
Vzorec: C₁₂H₁₆O₂
Hustota: 0,99 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00026515
CAS číslo: 94-46-2
EINECS: 202-334-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR