3-Hydroxybenzhydrazide ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-Hydroxybenzoic hydrazide, 3-Hydroxybenzoic acid hydrazide, 3-Hydroxybenzohydrazide, 3-Hydroxybenzene-1-carbohydrazide
L04357.06
L04357.03
L04357.06EA
3020
CZK
L04357.06
L04357.03
3-Hydroxybenzhydrazide ≥98%
3-Hydroxybenzhydrazide
Purity: 98+%
CAS No.: 5818-06-4
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Melting Point: 157-159°
Beilstein Reference: 638933
Vzorec:
C₇H₈N₂O₂ Bod tání: 157…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014759 CAS číslo: 5818-06-4 EINECS: 227-389-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...