o-Anisohydrazide ≥98%
Dodavatel: Thermo Scientific
Synonyms:
2-Methoxybenzoic hydrazide, 2-Methoxybenzhydrazide, 2-Methoxybenzohydrazide, 2-Methoxybenzoic acid hydrazide, 2-Methoxybenzene-1-carbohydrazide, 2-Anisohydrazide
A13619.09
A13619.18
A13619.09EA
3570
CZK
A13619.09
A13619.18
o-Anisohydrazide ≥98%
o-Anisohydrazide
Purity: 98+%
CAS No.: 7466-54-8
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Melting Point: 78-82°
Beilstein Reference: 2208367
Vzorec:
C₈H₁₀N₂O₂ MW: 166,18 g/mol Bod tání: 78…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014755 CAS číslo: 7466-54-8 EINECS: 231-260-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...