4-Nitrobenzhydrazide ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Nitrobenzoic hydrazide
A18016.09
A18016.18
A18016.09EA
2460
CZK
A18016.09
A18016.18
4-Nitrobenzhydrazide ≥98%
4-Nitrobenzhydrazide
Purity: 98+%
CAS No.: 636-97-5
Molecular Formula: C7H7N3O3
Formula Weight: 181.15
Melting Point: ca 216° dec.
Beilstein Reference: 519882
Vzorec:
C₇H₇N₃O₃ MW: 181,15 g/mol Bod tání: ∼ 216 °C (rozkl.) Storage Temperature: Ambient |
MDL Number:
MFCD00007604 CAS číslo: 636-97-5 EINECS: 211-271-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...