Anthranilohydrazide ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
2-Aminobenzoic hydrazide, 2-Aminobenzoic acid hydrazide, 2-Aminobenzohydrazide, 2-Aminobenzoylhydrazide, Anthraniloyl Hydrazine, 2-Aminobenzene-1-carbohydrazide, 2-Aminobenzhydrazide
A10805.09
A10805.18
A10805.09EA
1680
CZK
A10805.09
A10805.18
Anthranilohydrazide ≥98%
Anthranilohydrazide
Purity: 98+%
CAS No.: 1904-58-1
Molecular Formula: C7H9N3O
Formula Weight: 151.17
Melting Point: 122-124°
Beilstein Reference: 742811
Vzorec:
C₇H₉N₃O MW: 151,17 g/mol Bod varu: 378 °C (1013 hPa) Bod tání: 122…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017056 CAS číslo: 1904-58-1 EINECS: 217-604-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...