Tisk…

Benzene-1,3,5-tricarbonyl trichloride, Sigma-Aldrich®

Dodavatel: Merck
Nebezpečí


M-Clarity™ quality level = MQ200

147532-100G 147532-10G 147532-25G 147532-500G
SIAL147532-100GEA 12520 CZK
SIAL147532-100G SIAL147532-10G SIAL147532-25G SIAL147532-500G
Benzene-1,3,5-tricarbonyl trichloride, Sigma-Aldrich®
Benzene-1,3,5-tricarbonyl trichloride
Vzorec: C₉H₃Cl₃O₃
Bod varu: 312 °C (1013 hPa)
Bod tání: 34…36 °C
Storage Temperature: Ambient
MDL Number: MFCD00000679
CAS číslo: 4422-95-1
EINECS: 224-594-8
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR