3,5-Dimethoxybenzoyl chloride, Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
M-Clarity™ quality level = MQ100
161713-10G
161713-50G
SIAL161713-10GEA
5360
CZK
SIAL161713-10G
SIAL161713-50G
3,5-Dimethoxybenzoyl chloride, Sigma-Aldrich®
3,5-Dimethoxybenzoyl chloride
Vzorec:
C₉H₉ClO₃ MW: 200,62 g/mol Bod varu: 284 °C (1013 hPa) Bod tání: 35…36 °C |
MDL Number:
MFCD00000676 CAS číslo: 17213-57-9 EINECS: 241-256-5 UN: 3261 ADR: 8,II |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...