3-Anisoyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
3-Methoxybenzoyl chloride, m-Anisoyl chloride
M-Clarity™ quality level = MQ100
230243-25G
230243-5G
SIAL230243-25GEA
4670
CZK
SIAL230243-25G
SIAL230243-5G
3-Anisoyl chloride, Sigma-Aldrich®
3-Anisoyl chloride
Vzorec:
C₈H₇ClO₂ Bod varu: 123 °C (1013 hPa) Hustota: 1,21 g/cm³ (20 °C) |
MDL Number:
MFCD00000673 CAS číslo: 1711-05-3 EINECS: 216-975-2 UN: 1729 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...