Tisk…

4-(Dimethylamino)benzoyl chloride 97%

Dodavatel: Thermo Scientific
Nebezpečí

L14176.06 L14176.14
L14176.06EA 1450 CZK
L14176.06 L14176.14
4-(Dimethylamino)benzoyl chloride 97%
4-(Dimethylamino)benzoyl chloride

Purity: 97% CAS No.: 4755-50-4 Molecular Formula: C9H10ClNO Formula Weight: 183.64 Melting Point: 142-148° Beilstein Reference: 1100858 UN No.: 3261 MDL No.: MFCD00236039

Vzorec: C₉H₁₀ClNO
Bod varu: 180 °C (15 mmHg)
Bod tání: 142…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00236039
CAS číslo: 4755-50-4
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR