Tisk…

3-Acetamidobenzoic acid ≥98%

Dodavatel: Thermo Fisher Scientific
Varování

Synonyms: 3-Acetamidobenzoic Acid, 3-Acetylaminobenzoic acid

A15902.14 A15902.22
A15902.14EA 927 CZK
A15902.14 A15902.22
3-Acetamidobenzoic acid ≥98%
3-Acetamidobenzoic acid

Purity: 98% CAS No.: 587-48-4 Molecular Formula: C9H9NO3 Formula Weight: 179.18 Melting Point: 249-252° Beilstein Reference: 2210867

Vzorec: C₉H₉NO₃
MW: 179.18
Bod tání: 249…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00013983
CAS číslo: 587-48-4
EINECS: 209-600-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR