Terephthalonitrile 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1,4-Benzenedicarbonitrile, Terephthalodinitrile, Benzene-1,4-dicarbonitrile, 1,4-Dicyanobenzene
113820250
113821000
113825000
ACRO113820250EA
1140
CZK
ACRO113820250
ACRO113821000
ACRO113825000
Terephthalonitrile 98%
Terephthalonitrile
Vzorec:
C₈H₄N₂ MW: 128,13 g/mol Bod tání: 222…227 °C Hustota: 1,29 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00001810 CAS číslo: 623-26-7 EINECS: 210-783-2 |
Výsledky specifikačního testu
Appearance | White to light yellow to light beige Powder |
Infrared spectrum | Conforms |
Melting point | 223 to 230 °C |
HPLC | ≥97.5 % |
Ammonium (NH4) | ≤20 ppm |
Iron (Fe) | ≤2 ppm |
Sulfur (S) | ≤10 ppm |
Heavy metals (as Pb) | ≤5 ppm |
Impurity | ≤0.2 % Carbonate (as Na2CO3) |
Insoluble matter | ≤0.005 % |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...