4-Cyanobenzenesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
4-Cyanobenzene-1-sulphonyl chloride
M-Clarity™ quality level = MQ200
638331-1G
638331-5G
SIAL638331-1GEA
0
CZK
SIAL638331-1G
SIAL638331-5G
4-Cyanobenzenesulphonyl chloride, Sigma-Aldrich®
4-Cyanobenzenesulphonyl chloride
Vzorec:
C₇H₄ClNO₂S MW: 201,63 g/mol |
MDL Number:
MFCD00052337 CAS číslo: 49584-26-1 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...