Tisk…

3-Cyanobenzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3-Cyanobenzene-1-sulphonyl chloride, 3-Cyanobenzene-1-sulphonylchloride

M-Clarity™ quality level = MQ200

638358-1G 638358-5G
SIAL638358-1GEA 0 CZK
SIAL638358-1G SIAL638358-5G
3-Cyanobenzenesulphonyl chloride, Sigma-Aldrich®
3-Cyanobenzenesulphonyl chloride
Vzorec: C₇H₄ClNO₂S
MW: 201,63 g/mol
MDL Number: MFCD00203480
CAS číslo: 56542-67-7
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR