Tisk…

Methyl-4-cyanobenzoate, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Cyanobenzoic acid methyl ester

M-Clarity™ quality level = MQ100

230634-25G 230634-5G
SIAL230634-25GEA 6080 CZK
SIAL230634-25G SIAL230634-5G
Methyl-4-cyanobenzoate, Sigma-Aldrich®
Methyl-4-cyanobenzoate
Vzorec: C₉H₇NO₂
MW: 161,16 g/mol
Bod varu: 275 °C (1013 hPa)
Bod tání: 67 °C
MDL Number: MFCD00001823
CAS číslo: 1129-35-7
EINECS: 214-443-4
UN: 3439
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR