Tisk…

4-Chlorobenzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 4-Chlorobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

133698-100G 133698-500G 133698-5G
SIAL133698-100GEA 1660 CZK
SIAL133698-100G SIAL133698-500G SIAL133698-5G
4-Chlorobenzenesulphonyl chloride, Sigma-Aldrich®
4-Chlorobenzenesulphonyl chloride
Vzorec: C₆H₄Cl₂O₂S
MW: 211,07 g/mol
Bod varu: 272 °C (1013 hPa)
Bod tání: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00007439
CAS číslo: 98-60-2
EINECS: 202-685-3
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR