2,4,6-Trichlorobenzenesulphonyl chloride, Sigma-Aldrich®
Dodavatel: Merck
Synonyms:
2,4,6-Trichlorobenzene-1-sulphonyl chloride
M-Clarity™ quality level = MQ100
555649-1G
555649-5G
SIAL555649-1GEA
1340
CZK
SIAL555649-1G
SIAL555649-5G
2,4,6-Trichlorobenzenesulphonyl chloride, Sigma-Aldrich®
2,4,6-Trichlorobenzenesulphonyl chloride
Vzorec:
C₆H₂Cl₄O₂S MW: 279,96 g/mol Bod tání: 49 °C |
MDL Number:
MFCD00052973 CAS číslo: 51527-73-2 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...