Tisk…

2,4,5-Trichlorobenzenesulphonyl chloride, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 2,4,5-Trichlorobenzene-1-sulphonyl chloride

M-Clarity™ quality level = MQ100

680125-25G 680125-5G
SIAL680125-25GEA 5220 CZK
SIAL680125-25G SIAL680125-5G
2,4,5-Trichlorobenzenesulphonyl chloride, Sigma-Aldrich®
2,4,5-Trichlorobenzenesulphonyl chloride
Vzorec: C₆H₂Cl₄O₂S
MW: 279,96 g/mol
Bod tání: 69 °C
Hustota: 1,88 g/cm³ (20 °C)
MDL Number: MFCD00007428
CAS číslo: 15945-07-0
EINECS: 240-079-0
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR