Tisk…

2,4,5-Trichlorobenzenesulphonyl chloride ≥98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 2,4,5-Trichlorobenzene-1-sulphonyl chloride

A19390.06 A19390.22
A19390.06EA 951 CZK
A19390.06 A19390.22
2,4,5-Trichlorobenzenesulphonyl chloride ≥98%
2,4,5-Trichlorobenzenesulphonyl chloride

Purity: 98% CAS No.: 15945-07-0 Molecular Formula: C6H2Cl4O2S Formula Weight: 279.96 Melting Point: 67-71° Beilstein Reference: 1112595 UN No.: 3261 MDL No.: MFCD00007428

Vzorec: C₆H₂Cl₄O₂S
Bod tání: 67…71 °C
Hustota: 1,88 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007428
CAS číslo: 15945-07-0
EINECS: 240-079-0
UN: 3261
ADR: 8,II

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR