Tisk…

2-Chlorothiophenol, Sigma-Aldrich®

Dodavatel: Merck


M-Clarity™ quality level = MQ100

247146-25G 247146-5G
SIAL247146-25GEA 6720 CZK
SIAL247146-25G SIAL247146-5G
2-Chlorothiophenol, Sigma-Aldrich®
2-Chlorothiophenol
Vzorec: C₆H₅ClS
MDL Number: MFCD00004830
CAS číslo: 6320-03-2
EINECS: 228-667-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR