4-Chlorothioanisole ≥98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Chloro-4-(methylthio)benzene, 4-Chlorophenyl methyl sulphide
A11025.06
A11025.14
A11025.06EA
1270
CZK
A11025.06
A11025.14
4-Chlorothioanisole ≥98%
4-Chlorothioanisole
Purity: 98%
CAS No.: 123-09-1
Molecular Formula: C7H7ClS
Formula Weight: 158.65
Flash Point: 90°(194°F)
Refractive Index: 1.600
Odour: Unpleasant
Boiling Point: 223°
Beilstein Reference: 2041509
MDL No.: MFCD00013643
Vzorec:
C₇H₇ClS MW: 158,65 g/mol Bod varu: 223 °C Bod tání: 27…28 °C Hustota: 1,220 Bod vzplanutí: 90 °C (194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00013643 CAS číslo: 123-09-1 EINECS: 204-600-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...