4-Chlororesorcinol ≥98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Chlorobenzene-1,3-diol, 1-Chloro-2,4-dihydroxybenzene
B24076.14
B24076.22
B24076.36
B24076.14EA
858
CZK
B24076.14
B24076.22
B24076.36
4-Chlororesorcinol ≥98%
4-Chlororesorcinol
Purity: 98%
CAS No.: 95-88-5
Molecular Formula: C6H5ClO2
Formula Weight: 144.56
Melting Point: 108-111°
Boiling Point: 146-148°/18mm
MDL No.: MFCD00002273
Vzorec:
C₆H₅ClO₂ Bod varu: 146…148 °C (18 mmHg) Bod tání: 108…111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002273 CAS číslo: 95-88-5 EINECS: 202-462-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...