Tisk…

4-Chloroanisole, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 1-Chloro-4-methoxybenzene

M-Clarity™ quality level = MQ100

159069-100G 159069-25G
SIAL159069-100GEA 7030 CZK
SIAL159069-100G SIAL159069-25G
4-Chloroanisole, Sigma-Aldrich®
4-Chloroanisole
Vzorec: C₇H₇ClO
MW: 142,58 g/mol
Bod varu: 200 °C (1013 hPa)
Bod tání: –18 °C
Hustota: 1,17 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000616
CAS číslo: 623-12-1
EINECS: 210-772-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR