4-Amino-2,6-dichlorophenol 97%
Dodavatel: Thermo Scientific
|
Varování
|
A11568.09
A11568.18
A11568.30
A11568.09EA
937
CZK
A11568.09
A11568.18
A11568.30
4-Amino-2,6-dichlorophenol 97%
4-Amino-2,6-dichlorophenol
Purity: 97%
CAS No.: 5930-28-9
Molecular Formula: C6H5Cl2NO
Formula Weight: 178.02
Melting Point: 164-169°
Beilstein Reference: 2361665
Vzorec:
C₆H₅Cl₂NO MW: 178,02 g/mol Bod varu: 304 °C (1013 hPa) Bod tání: 164…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007875 CAS číslo: 5930-28-9 EINECS: 227-671-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...