Tisk…

5-Chloro-2-nitroaniline ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

A11241.06 A11241.14
A11241.06EA 1110 CZK
A11241.06 A11241.14
5-Chloro-2-nitroaniline ≥97%
5-Chloro-2-nitroaniline

Purity: 97% CAS No.: 1635-61-6 Molecular Formula: C6H5ClN2O2 Formula Weight: 172.57 Odour: Weak, characteristic Melting Point: 124-128° Beilstein Reference: 2210201 UN No.: 2237 MDL No.: MFCD00007776

Vzorec: C₆H₅ClN₂O₂
MW: 172,57 g/mol
Bod varu: 323 °C (1013 hPa)
Bod tání: 124…128 °C
Storage Temperature: Ambient
MDL Number: MFCD00007776
CAS číslo: 1635-61-6
EINECS: 216-661-5
UN: 2237
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR