Dicloran (2,6-dichloro-4-nitroaniline), Sigma-Aldrich®
Dodavatel: Merck
|
Nebezpečí
|
Synonyms:
2,6-Dichloro-4-nitroaniline
M-Clarity™ quality level = MQ200
D67820-100G
D67820-5G
SIALD67820-100GEA
1230
CZK
SIALD67820-100G
SIALD67820-5G
Dicloran (2,6-dichloro-4-nitroaniline), Sigma-Aldrich®
Dicloran (2,6-dichloro-4-nitroaniline)
Vzorec:
C₆H₄Cl₂N₂O₂ Bod varu: 323 °C (1013 hPa) Bod tání: 186…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007677 CAS číslo: 99-30-9 EINECS: 202-746-4 UN: 2237 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...