(3-Chlorfenyl)isokyanát, Sigma-Aldrich®
Dodavatel: Merck
M-Clarity™ quality level = MQ100
245682-100G
245682-5G
SIAL245682-100GEA
1370
CZK
SIAL245682-100G
SIAL245682-5G
(3-Chlorfenyl)isokyanát, Sigma-Aldrich®
(3-Chlorfenyl)isokyanát
Vzorec:
C₇H₄ClNO Bod varu: 202 °C (1013 hPa) Hustota: 1,26 g/cm³ (20 °C) |
MDL Number:
MFCD00002016 CAS číslo: 2909-38-8 EINECS: 220-822-5 UN: 3382 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...