Tisk…

3,4-Dichlorophenyl isocyanate ≥97%

Dodavatel: Thermo Scientific
Nebezpečí

A16298.06 A16298.14
A16298.06EA 1490 CZK
A16298.06 A16298.14
3,4-Dichlorophenyl isocyanate ≥97%
3,4-Dichlorophenyl isocyanate

Purity: 97% CAS No.: 102-36-3 Molecular Formula: C7H3Cl2NO Formula Weight: 188.01 Flash Point: 123°(253°F) Odour: Irritating Boiling Point: 118-120°/18mm Beilstein Reference: 608325 UN No.: 2250 MDL No.: MFCD00002017

Vzorec: C₇H₃Cl₂NO
MW: 188,01 g/mol
Bod varu: 118…120 °C (18 mmHg)
Bod tání: 40…44 °C
Bod vzplanutí: 123 °C (253 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002017
CAS číslo: 102-36-3
EINECS: 203-026-2
UN: 2250
ADR: 6.1,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR