2-Chloro-5-iodobenzoic acid ≥97%
Dodavatel: Thermo Scientific
|
Varování
|
L20161.06
L20161.14
L20161.22
L20161.06EA
1990
CZK
L20161.06
L20161.14
L20161.22
2-Chloro-5-iodobenzoic acid ≥97%
2-Chloro-5-iodobenzoic acid
Purity: 97%
CAS No.: 19094-56-5
Molecular Formula: C7H4ClIO2
Formula Weight: 282.46
Melting Point: 156-161°
Beilstein Reference: 2254110
Vzorec:
C₇H₄ClIO₂ MW: 282,46 g/mol Bod tání: 156…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00079731 CAS číslo: 19094-56-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...