Tisk…

1,3-Dichloro-5-iodobenzene, Sigma-Aldrich®

Dodavatel: Merck

Synonyms: 3,5-Dichloroiodobenzene

M-Clarity™ quality level = MQ100

192546-25G 192546-5G
SIAL192546-25GEA 6090 CZK
SIAL192546-25G SIAL192546-5G
1,3-Dichloro-5-iodobenzene, Sigma-Aldrich®
1,3-Dichloro-5-iodobenzene
Vzorec: C₆H₃Cl₂I
MW: 272,9 g/mol
Bod varu: 127 °C (20 mmHg)
Bod tání: 57 °C
MDL Number: MFCD00001047
CAS číslo: 3032-81-3
EINECS: 221-216-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR