Tisk…

2-Chloro-6-fluoroaniline ≥98%

Dodavatel: Thermo Scientific
Nebezpečí

A15848.03 A15848.06 A15848.14
A15848.03EA 1270 CZK
A15848.03 A15848.06 A15848.14
2-Chloro-6-fluoroaniline ≥98%
2-Chloro-6-fluoroaniline

Purity: 98% CAS No.: 363-51-9 Molecular Formula: C6H5ClFN Formula Weight: 145.56 Boiling Point: 67-69°/14mm UN No.: 2810 MDL No.: MFCD00040309

Vzorec: C₆H₅ClFN
MW: 145,56 g/mol
Bod varu: 67…69 °C (14 mmHg)
Bod tání: 31…32 °C
Hustota: 1,316
Storage Temperature: Ambient
MDL Number: MFCD00040309
CAS číslo: 363-51-9
EINECS: 206-657-1
UN: 2810
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR