Tisk…

2-Chloro-6-fluorobenzamide ≥98%

Dodavatel: Thermo Scientific
Varování

B21294.06 B21294.14
B21294.06EA 1010 CZK
B21294.06 B21294.14
2-Chloro-6-fluorobenzamide ≥98%
2-Chloro-6-fluorobenzamide

Purity: 98% CAS No.: 66073-54-9 Molecular Formula: C7H5ClFNO Formula Weight: 173.58 Melting Point: 139-142° MDL No.: MFCD00055436

Vzorec: C₇H₅ClFNO
Bod tání: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00055436
CAS číslo: 66073-54-9
EINECS: 266-123-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR